Check Order Status
Cotton Canvas Gloves
Actual product may vary
Cotton Canvas Gloves
Multi-purpose fabric gloves used for light protection in many general applications.
Have Questions?
Change view
Catalog Number
Gender
FIS19-013526
Women's
FIS19-013522
Men's
2 Options
Catalog number
FIS19-013526
Price (SGD)
29.00
PK
Add to cart
Gender:
Women's
Price (SGD)
29.00
PK
Add to cart
Heavyweight fabric provides increased protection
Knitwrist prevents dirt and debris from entering the glove
Blended cotton/polyester fabric
Clute cut
Specifications
Color
Natural
Grade
Economy
Material
Cotton, Canvas
Product Type
General Purpose Gloves
Style
Clute
For Use With (Application)
Light protection in general applications
Gender
Women's
Glove Cuff Style
Knit