Extra Heavyweight Terry Cloth Heat Protection Gloves - Gauntlet Cuff
Actual product may vary
Wells Lamont
Extra Heavyweight Terry Cloth Heat Protection Gloves - Gauntlet Cuff
Extra Heavyweight Terry Cloth Heat Protection Gloves are constructed of extra heavyweight terry cloth with a 5 in. gauntlet cuff for protection from temperatures up to 450°F.
Have Questions?
Catalog NumberSize
FIS19-013299X-Large
Catalog number FIS19-013299
Price (SGD)
359.00
Dozen
Size:
X-Large
Price (SGD)
359.00
Dozen
  • Extra heavy weight terry cloth provides protection from temperatures up to 450°F
  • 5 in. gauntlet cuff helps protect the arm from burns
  • Loop-out construction
  • Ambidextrous
Specifications
ColorBrown
HandAmbidextrous
MaterialTerrycloth
Product TypeHeat Protection Gloves
SizeX-Large
Temperature (English) Max.450°F
Temperature (Metric) Max232°C
For Use With (Application)Casting, Stamping, and Steel Processing
Glove Cuff StyleGauntlet
Series636HRL
Length (English) Cuff5 in.
Length (Metric) Cuff12.7 cm
Unit SizeDozen