Meta 6116 Unisex 40 Inch Lab Coats
Actual product may vary
White Swan
Meta 6116 Unisex 40 Inch Lab Coats
Long length lab coat features four pockets (one inside), hand access slits, and a two-piece back swing belt.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSize
FIS18-999-942FWhite2X-Large
FIS18-999-942CWhiteMedium
FIS18-999-942BWhiteSmall
FIS18-999-942DWhiteLarge
FIS18-999-942EWhiteX-Large
FIS18-999-944CNavyMedium
FIS18-999-944GNavy3X-Large
Catalog number FIS18-999-942F
Price (SGD)
-
Color:
White
Size:
2X-Large
  • Not fluid resistant or antimicrobial
  • 4 pockets
  • Slits provide access to pants pockets
  • Made of 65% polyester, 35% cotton poplin
  • Three patch-style pockets on outside, plus one inside pocket
  • Button-front closure
  • Two-piece swing belt
  • Length: 40 in. (102 cm)
  • Washable
  • Tall and sizes up to 4X available upon request
  • Embroidery available upon request
Specifications
Product TypeGeneral Purpose Lab Coat
Size2X-Large
GenderUnisex
MaterialPolycotton
ColorWhite
Closure TypeButtons
DisposableReusable
SterilityNon-sterile
StyleLong
No. of Pockets4
FinishSoil Release
Size (English) Chest50 to 52 in.
Size (English) Waist44 to 46 in.
Size (Metric) Waist111.7 to 116.8 cm
Length (English)40 in.
Length (Metric)102 cm
ConstructionPolyester, Cotton Poplin, Three Patch Style Pockets Outside, One Inside Pocket
Unit SizeEach