Hazardous Materials Disposal Bags
Actual product may vary
Hazardous Materials Disposal Bags
Hazardous Materials Disposal Bags
New Pig
Hazardous Materials Disposal Bags
Confidently bag hazardous or contaminated waste for disposal.
Have Questions?
Catalog NumberCapacity (English)
FIS18-999-570145 gal.
Catalog number FIS18-999-5701
Price (SGD)
286.00
Case
Capacity (English):
45 gal.
Price (SGD)
286.00
Case
  • Tough polyethylene bags resist punctures, tearing and blowouts
  • Durable 3 mil thickness
  • Bright yellow bags are boldly labeled Hazardous Material - Handle With Care
Specifications
Capacity (Metric)170.3 L
Capacity (English)45 gal.
ColorYellow, Black
Height (Metric)152.4 cm
Width (Metric)91.44 cm
For Use With (Application)Ideal for lining 30 and 55 gal. drums or for temporary storage of saturated absorbents and other waste
Width (English)36 in.
Thickness3 mil
Height (English)60 in.
MaterialPolyethylene
Product TypeHazardous Disposal Bag
Unit SizeCase