M3 Pleat-Style Face Masks
Actual product may vary
Ansell
M3 Pleat-Style Face Masks
Pleat-style mask available with earloops or soft ties. Suggested for use in ISO Class 3 and higher cleanroom environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFastening TypeWidth (Metric)Width (English)
FIS18-999-4775Ties18 cm7.1 in.
FIS18-999-4774Earloops18 cm7.1 in.
FKS11742473Earloops18 cm7.1 in.
Catalog number FIS18-999-4775
Price (SGD)
669.00
Case
Fastening Type:
Ties
Width (Metric):
18 cm
Width (English):
7.1 in.
Price (SGD)
669.00
Case
  • Inner facing made of white BiCoSof™
  • Nose piece made from fully enclosed, soft, malleable aluminum
  • Outer facing made from clear polyethylene apertured film
Specifications
Product TypeCleanroom Face Mask
ColorWhite
Fastening TypeTies
MaterialPolyethylene
SizeStandard
SterilityNon-sterile
StylePleated
Cleanroom ClassClass 1 (ISO 3)
DisposableSingle-use
Filter Efficiency99.5% (PFE), 98.7% (BFE)
For Use With (Application)Semiconductor, Disk Drive, Data Storage, Integrated Circuits
Latex FreeYes
Model62466
Pore Size0.1 μm (PFE), 3 μm (BFE)
SeriesKimtech™ M3
Width (English)7.1 in.
Width (Metric)18 cm
Unit SizeCase