Single-Drum Spill Containment Unit
Actual product may vary
Eagle
Single-Drum Spill Containment Unit
Suitable for forklift use
Have Questions?
Catalog NumberCapacity (English)
FIS18-999-447365 gal.
Catalog number FIS18-999-4473
Price (SGD)
677.00
Each
Capacity (English):
65 gal.
Price (SGD)
677.00
Each
Ideal for use as spill containment unit, mobile pumping station, or waste collection station.
  • Provides secondary spill containment of hazardous materials when used with drums up to 55 gal. (208L)
  • Constructed of high-density polyethylene with UV inhibitors for superior strength and excellent chemical resistance
  • Nestable for easy storage
Specifications
Certifications/ComplianceEPA, 40 CFR 264.175
ColorYellow
Capacity (English)65 gal.
Capacity (Metric)246 L
MaterialHDPE
TypeSingle Drum Containment Unit
For Use With (Application)Forklift use
For Use With (Equipment)For use as spill containment unit, mobile pumping station, or waste collection station
Holds1 Drum
Includes0.75 in. drain plug, black grating with 1.75 in. openings, 78.7 x 83.8cm
ResistantChemical
Length (English)31 in.
Length (Metric)78.7 cm
Height (English)33 in.
Height (Metric)83.8 cm
Weight (English)30 lb.
Weight (Metric)13.6 kg
Unit SizeEach