Splash Hood
Actual product may vary
Oberon
Splash Hood
Full head protection from intermittent liquid splash
Have Questions?
Catalog number FIS18-999-2641
Price (SGD)
-
  • Shell is designed with a full shape to protect the head, neck and shoulders
  • Chemical-resistant coating on outer surface of window matches chemical/splash resistance of vinyl coated fabric
  • Face-Fit™ chemical-resistant polycarbonate Window coated with Silicote™-6 resists damage caused by many mild acids, bases and solvents (except hydrofluoric acid)
  • Inside of the window is permanent antifog coated for clear visibility and no fogging
  • Includes ratchet headgear for easy adjustment
  • Made in the USA
Will not wipe off hydrofluoric acid
Specifications
Lens CoatingAnti-fog
Certifications/ComplianceANSI Z87.1-2010
Product TypeSplash Hood
IncludesVinyl coated fabric hood; Polycarbonate window
MaterialVinyl
Lens MaterialPolycarbonate
Unit SizeEach