Kimtech™ M3 Sterile Face Masks
Actual product may vary
Ansell
Kimtech™ M3 Sterile Face Masks
Sterile masks feature non-irritating BiCoSof™ inner facing and bendable aluminum nosepiece. Available with earloops or soft ties.
Have Questions?
Catalog NumberFastening TypeIncludes
FIS18-999-2405Ties7 in. Soft Ties
Catalog number FIS18-999-2405
Price (SGD)
831.00
Case
Fastening Type:
Ties
Includes:
7 in. Soft Ties
Price (SGD)
831.00
Case
Sterile masks feature non-irritating BiCoSof™ inner facing and bendable aluminum nosepiece. Available with earloops or soft ties.
  • Gamma-sterilized to 10-6
  • White with soft ties or earloops
  • Outer layer of apertured polyethylene film to provide protection from contamination
  • Polyester binding and polyester filter media provide bacteria filtration efficiency of 91.9% and particle filtration efficiency of 93.1%
Specifications
Product TypeCleanroom Face Mask
ColorWhite
Fastening TypeTies
MaterialSMS
SizeStandard
SterilitySterile
StylePleated
Cleanroom ClassClass 1 (ISO 3)
DisposableSingle-use
Filter Efficiency89.3% (PFE), 94.1% (BFE)
Includes7 in. Soft Ties
Latex FreeYes
Model62467
Pore Size0.1 μm (PFE), 3 μm (BFE)
SeriesKimtech™ M3 Sterile
Unit SizeCase