SparkPlugs™ Disposable Ear Plugs
Actual product may vary
Moldex
SparkPlugs™ Disposable Ear Plugs
Multi-colored SparkPlugs™ ear plugs are made of extra-soft, extra light foam. Tapered shape fits easily in the ear canal to seal gently and comfortably.
Have Questions?
Catalog NumberCorded
FIS18-999-2271Uncorded
Catalog number FIS18-999-2271
Price (SGD)
131.00
Pack
Corded:
Uncorded
Price (SGD)
131.00
Pack
  • Extra-soft, extra-light foam with nonirritating surface
  • Tapered for easy insertion and removal
  • Seals ear canal comfortably, without pressure
  • Disposable
  • Every package reveals random multi-colored SparkPlugs
  • Provide a secure fit in all sizes of ear canals
  • Hygienically packaged
  • 100% PVC-Free™
Specifications
ANSI S3.1974Yes
CSA Z94.2Class AL
SizeUniversal
For Use With (Application)Construction, Energy, Utilities, Manufacturing, Healthcare, Mining, Oil and Gas, Transportation, and Other Various Industries
High VisibilityNo
Packaging TypePoly bag
Pairs per Package200 Pairs/Pk.
Plug MaterialFoam
Plug ColorMulti-color
Noise Reduction Rating (NRR)33 dB
CordedUncorded
Plug ShapeBullet
DisposableSingle-use
Metal DetectableNo
Unit SizePack