Overpack Dolly
Actual product may vary
Eagle
Overpack Dolly
Safely moves salvage drums
Have Questions?
Catalog NumberQuantity
FIS18-869
Catalog number FIS18-869
Price (SGD)
-
  • Heavy-duty dolly holds 85 gal. (323L) and 95 gal. (361L) overpack salvage drums
  • Welded construction with rubber wheels
Specifications
Height (English)7 in.
Product TypeDrum Dolly
ColorBlack
Height (Metric)17.7 cm
Unit SizeEach