PELTOR™ Optime™ 105 Ear Muffs
PELTOR™ Optime™ 105 Ear Muffs
Actual product may vary
Fisherbrand
PELTOR™ Optime™ 105 Ear Muffs
Optime™ Ear Muffs provide maximum sound quality in the most demanding environments where noise exposures are expected to reach as high as 105 dBA.
Have Questions?
Catalog NumberModelColorStyle
FIS18-858MH10ARedOver-the-head
Catalog number FIS18-858M
Price (SGD)
130.00
EA
Add to cart
Model:
H10A
Color:
Red
Style:
Over-the-head
Price (SGD)
130.00
EA
Add to cart
  • Effective hearing protection for noise exposures up to 105dBA
  • Double-shell ear cup design technology boosts noise blocking ability
  • Soft, foam-filled ear cushions provide added comfort
  • Durable stainless steel headband resists bending and warping
  • Available in headband style red/black or high-visibility green color
  • Available in neckband and hard hat-attached styles
Specifications
ColorRed
ANSI S3.1974Yes
Cushion MaterialFoam
CSA Z94.2Class AL
Cushion ColorBlack
Headband MaterialStainless Steel
For Use With (Application)Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
ModelH10A
Pairs per Package1 Ea.
Noise Reduction Rating (NRR)30 dB
Product TypePELTOR™ Optime 105
Headband ColorBlack
DielectricNo
MaterialABS
StyleOver-the-head
Unit SizeEA