Colored Delineator Tape
Actual product may vary
Harris Industries
Colored Delineator Tape
Designed for safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities.
Have Questions?
Catalog NumberColorDimensions (L x W)
FIS18-785Red, White36 yd. x 2 in.
Catalog number FIS18-785
Price (SGD)
-
Color:
Red, White
Dimensions (L x W):
36 yd. x 2 in.
Features:
  • Alternating color stripes
  • Highly visible
  • Pressure sensitive
  • Rubber-based permanent adhesive
  • Self-wound
  • Clear laminate resists oil, dirt, chemicals and abrasion
  • 6.0mil/ 0.150mm thickness
Benefits:
  • Laminated PVC will not fade or scuff
  • Holds up to traffic and wear longer than paint
  • Can be applied quickly and easily with minimal surface preparation
  • Maintenance-free; brilliance is restored with wet mopping/washing
Red/White: Fire Protection Equipment; Black/Yellow: Physical Hazard; Black/White: Housekeeping Aisle Marking
Specifications
Certifications/ComplianceOSHA 1910.144
Dimensions (L x W)36 yd. x 2 in.
For Use With (Application)Safety marking applications in industrial plants, commercial buildings, institutions and recreational facilities, resistance to oil, dirt and chemicals
Height (English)2 in.
Height (Metric)5.08 cm
Length (English)36 yd.
Length (Metric)32.9 m
Shelf Life1 Year
Temperature Range (English)14°F to 176°F
Temperature Range (Metric)-10°C to +80°C
Tensile Strength23N/cm
Thickness6 mil
Width (English)2 in.
Width (Metric)5.08 cm
Adhesive TypeRubber-based Adhesive
ColorRed, White
MaterialPVC
TypeHazard Warning Tape
Unit SizeEach