Hughes Shower Tester
Hughes Shower Tester
Actual product may vary
Hughes Shower Tester
Includes everything needed to flow-test overhead emergency showers
Have Questions?
Catalog NumberIncludes
FIS18-563Dust Cover Lid, Log to Record Test Results
Catalog number FIS18-563
Price (SGD)
432.00
EA
Add to cart
Includes:
Dust Cover Lid, Log to Record Test Results
Price (SGD)
432.00
EA
Add to cart
Includes everything needed to flow-test overhead emergency showers
  • Three-piece PVC plastic handle quickly assembles to support 11 in. (27.9cm) PVC support ring
  • Polylaminated Tyvek™ sleeve directs shower discharge into polyethylene bucket
Specifications
IncludesDust Cover Lid, Log to Record Test Results
MaterialPVC Plastic, Taffeta
Product TypeShower Tester
Unit SizeEA