CleanTip™ Swabs: CleanFoam™ Series
CleanTip™ Swabs: CleanFoam™ Series
Actual product may vary
CleanTip™ Swabs: CleanFoam™ Series
Excellent sorbency
Have Questions?
Catalog NumberLength (English)Tip StyleHandle Type
FIS18-3742.795 in.RigidCompact
Catalog number FIS18-374
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (English):
2.795 in.
Tip Style:
Rigid
Handle Type:
Compact
Excellent sorbency
Laundered CleanFoam heads constructed of 100 pores per inch open-cell polyurethane foam for excellent capacity and cushioning.
  • Cleanest foam swabs available with superior solvent capacity and contamination capture
  • Solvent-processed for very low levels of nonvolatile residue and particle generation
  • Good chemical resistance—can be used with solvents such as isopropyl alcohol and applying lubricants and other liquids
  • Heads thermally welded onto polypropylene handles, without any contaminating adhesivesConstructed from the cleanest materials available
  • High-precision automated process guarantees consistent performance, sizing and cleanliness
  • Silicone-free Bag-Within-A-Bag™ packaging consists of a solvent-resistant, thermally sealed exterior bag that protects the inner bags containing the swabs
  • Each outer bag is individually lot-coded for easy traceability and quality control
Specifications
ConstructionRigid head core, compact handle
Diameter (English) Handle0.118 in.
Length (English)2.795 in.
For Use With (Application)Applying and removing lubricants, adhesives and other solutions in a critical clean environment, scrubbing recessed areas, removal of excess materials, debris, cleaning intersecting surfaces and joints
Handle ColorLight Green
Product TypeSwab
Diameter (Metric) Handle3 mm
Length (Metric)71 mm
Length (English) Handle2.32 in.
Length (Metric) Handle59 mm
MaterialCleanFoam
SizeSmall
Thickness (Metric)3.5 mm
Length (English) Tip0.472 in.
Handle TypeCompact
Length (Metric) Tip12 mm
Width (Metric) Tip3.4 mm
Width (English) Tip0.13 in.
Diameter (Metric) Tip3.6 mm
Diameter (English) Tip0.142 in.
Adhesive TypeThermal
Handle MaterialPolypropylene
Tip StyleRigid
Tip MaterialPolyurethane Foam
Unit SizePK