TexWrite™ 10 MP Documentation Paper
Actual product may vary
Fisherbrand
TexWrite™ 10 MP Documentation Paper
100% synthetic sheet
Have Questions?
Catalog NumberColor
FIS18-339White, Blue
Catalog number FIS18-339
Price (SGD)
-
Color:
White, Blue
High-performance documentation material designed for ultraclean manufacturing environments that cannot tolerate cellulose contamination.
  • Ultraclean with low particle generation and toner shedding
  • Excellent ink capture with no risk of smearing
  • Low ESD potential for reduced risk of electrostatic damage
  • Compatible with inkjet, laser and toner-based printers and photocopiers
  • Ballpoint pens glide easily over surface without skips or jumps
  • Recyclable as a plastic
  • Autoclavable for use in sterile environments
Specifications
MaterialSynthetic
ColorWhite, Blue
Dimensions (L x W)8.5 x 11 in.
Length (English)8.5 in.
Length (Metric)216 mm
Width (English)11 in.
Width (Metric)279 mm
AutoclavableAutoclavable
For Use With (Application)Ultraclean manufacturing environments that cannot tolerate cellulose contamination
Product TypeCleanroom Documentation Paper
Unit SizeCase