TechniCloth™ II Blended Wipers
TechniCloth™ II Blended Wipers
Actual product may vary
TechniCloth™ II Blended Wipers
For ISO Class 6 and 7 applications
Have Questions?
Catalog NumberDimensions (L x W)
FIS18-330A9 x 9 in. (22.86 x 22.86 cm)
Catalog number FIS18-330A
Price (SGD)
1,375.00
EA
Add to cart
Dimensions (L x W):
9 x 9 in. (22.86 x 22.86 cm)
Price (SGD)
1,375.00
EA
Add to cart
  • Made of a hydroentangled blend of 55% cellulose and 45% polyester and treated with ITW Texwipe proprietary ultralow particulate (ULP) finish
  • Significantly reduces particulate generation
  • ULP finish formulated to resist extraction in commonly used solvents
Specifications
Product TypeDry Wipe
Length (English)9 in.
Length (Metric)22.9 cm
Width (English)9 in.
Width (Metric)22.9 cm
MaterialPolyester Cellulose
Basis Weight67
Certifications/ComplianceISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
Dimensions (L x W)9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application)Designed for critical environments where low particle generation and high absorbency are prime considerations, excellent for use in facilities for integrated circuit, printed circuit board and electronic components fabrications
Unit SizeEA