Alpha™ 10 Sealed-Border Wipers
Alpha™ 10 Sealed-Border Wipers
Actual product may vary
Alpha™ 10 Sealed-Border Wipers
For ISO Class 3 to 4 applications
Have Questions?
Catalog NumberDimensions (L x W)Product TypeSaturation Solution
FIS18-323B12 x 12 in.Dry WipeDry Wiper
Catalog number FIS18-323B
Price (SGD)
-
Dimensions (L x W):
12 x 12 in.
Product Type:
Dry Wipe
Saturation Solution:
Dry Wiper
  • Vectra™ processed—hands-free, continuous manufacturing purification and inspection
  • Double-knit, no-run interlock construction of 100% continuous-filament polyester
  • Patented sealed-border wiper for lowest releasable fibers and particles
  • Optimum balance of wiping efficiency, abrasion resistance, and sorbency
  • Ideal wiper for use in multiple environments
  • Lot-to-lot traceability
Specifications
Product TypeDry Wipe
Length (English)12 in.
Length (Metric)30.5 cm
Width (English)12 in.
Width (Metric)30.5 cm
MaterialPolyester
Basis Weight135
Certifications/ComplianceISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
ConstructionDouble-knit
EdgesSealed
Dimensions (L x W)12 x 12 in.
For Use With (Application)Ideal wipes for use in multiple environments
Saturation SolutionDry Wiper
Unit SizePK