All-Purpose Cotton Wipers
All-Purpose Cotton Wipers
Actual product may vary
All-Purpose Cotton Wipers
For ISO Class 7 applications
Have Questions?
Catalog NumberDimensions (L x W)
FIS18-308A4 x 4 in. (10.16 x 10.16 cm)
Catalog number FIS18-308A
Price (SGD)
526.00
PK
Add to cart
Dimensions (L x W):
4 x 4 in. (10.16 x 10.16 cm)
Price (SGD)
526.00
PK
Add to cart
For ISO Class 7 applications
  • Strong, highly absorbent 100% cotton wipers woven from high-quality, long-staple cotton fibers with tight-weave construction
  • Twill-patterned surface on both sides of the wiper traps and removes surface contaminants from the cleaned surface
  • Inherent antistatic characteristics at 40-60% relative humidity
  • Exceptional absorbency, strength, chemical and heat resistance and uniform surface texture make these wipers suitable for a wide range of critical applications
  • Bias cut at a 45° angle to ensure that edges will resist fraying and release of fibers
  • Autoclavable
Specifications
Product TypeCleanroom Wiper
Length (English)4 in.
Length (Metric)10.2 cm
Width (English)4 in.
Width (Metric)10.16 cm
MaterialCotton
AutoclavableAutoclavable
Basis Weight184
Certifications/ComplianceISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
Dimensions (L x W)4 x 4 in. (10.16 x 10.16 cm)
For Use With (Application)Cleaning, polishing and burnishing of Metallic and nonmetallic magnetic media disk surfaces
Unit SizePK