▸
Primary Antibodies
▸
All Primary Antibodies
▸
CCL6/C10 Antibody (RM0183-7J13), Allophycocyanin/Cy7, Novus Biologicals™
Actual product may vary
LevGo
CCL6/C10 Antibody (RM0183-7J13), Allophycocyanin/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Have Questions?
Catalog Number
Quantity
FIS18-001-029
0.1 mL
Catalog number
FIS18-001-029
Price (SGD)
24.00
Pack
Add to cart
Quantity:
0.1 mL
Price (SGD)
24.00
Pack
Add to cart
CCL6/C10 Monoclonal specifically detects CCL6/C10 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Antigen
CCL6/C10
Applications
Flow Cytometry
Classification
Monoclonal
Clone
RM0183-7J13
Conjugate
APC-Cyanine7
Dilution
Flow Cytometry
Formulation
PBS with 0.05% Sodium Azide
Gene Symbols
Ccl6
Host Species
Rat
Immunogen
Mouse recombinant CCL6/C10
Purification Method
Protein A or G purified
Quantity
0.1 mL
Research Discipline
Chemokines and Cytokines
Primary or Secondary
Primary
Gene ID (Entrez)
20305.0
Target Species
Mouse
Isotype
IgG2
Unit Size
Pack
Contents & Storage
Store at 4°C in the dark. Do not freeze.