Eco-smartFunnel™ Disposable Eco Friendly Paper Lab Funnel
Actual product may vary
Eco-smartFunnel™ Disposable Eco Friendly Paper Lab Funnel
Eco-smartFunnel™ Disposable Eco Friendly Paper Lab Funnel
Eco-smartFunnel™ Disposable Eco Friendly Paper Lab Funnel
Eco-smartFunnel™ Disposable Eco Friendly Paper Lab Funnel
LevGo
Eco-smartFunnel™ Disposable Eco Friendly Paper Lab Funnel
Eco-smartFunnel is a simple, cost-effective solution to reduce plastic use in the lab and reduce cross contamination from reuse of washed funnels
Have Questions?
Catalog NumberDimensions (L x W)
FIS18-001-0235 x 2.25 in. (12.7 x 5.7 mm)
Catalog number FIS18-001-023
Price (SGD)
50.00
Pack
Dimensions (L x W):
5 x 2.25 in. (12.7 x 5.7 mm)
Price (SGD)
50.00
Pack
  • Reduce cross contamination
  • Fold flat design uses less storage space
  • Great for powders and cold liquids
  • Can be used in Saliva Collection Applications
  • Eco friendly, biodegradable
  • Made in the USA
Specifications
DescriptionPaper Disposable Funnel
DisposableSingle-use
MaterialFSC Certified Food grade clay coated paperboard
For Use With (Application)Sample collection, pouring viscous liquids and powders
ShapeSquare
TypeDisposable Eco Funnel
Dimensions (L x W)5 x 2.25 in. (12.7 x 5.7 mm)
Weight (Metric)5 g
Unit SizePack