Check Order Status
Filter Disc with 10.5 mm Bore, Ground Cylindrical
Actual product may vary
Thermo Scientific
Filter Disc with 10.5 mm Bore, Ground Cylindrical
Filter Disc for use with gas distribution tubes and washing bottle heads.
Have Questions?
Catalog Number
Porosity
FIS17020
0
Catalog number
FIS17020
Price (SGD)
417.00
EA
Add to cart
Porosity:
0
Price (SGD)
417.00
EA
Add to cart
Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Material
Borosilicate Glass (3.3)
Sterility
Non-sterile
Includes
Bore, Ø 10.5 mm
Porosity
0
Thickness (Metric)
3.5 ± 0.8 mm
Diameter (Metric)
30 mm