Tyvek™ IC668B IsoClean™ Full Face Opening Hoods
Actual product may vary
Tyvek™ IC668B IsoClean™ Full Face Opening Hoods
Tyvek™ IC668B IsoClean™ Full Face Opening Hoods
DuPont
Tyvek™ IC668B IsoClean™ Full Face Opening Hoods
IsoClean open face hoods provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Available in non-sterile, clean-processed, and sterile versions.
Have Questions?
Catalog NumberHood TypeSterility
FIS17-988-113Clean and SterileSterile
Catalog number FIS17-988-113
Price (SGD)
1,675.00
Case
Hood Type:
Clean and Sterile
Sterility:
Sterile
Price (SGD)
1,675.00
Case
  • Bound seams are covered with garment fabric for reinforcement
  • Bound hood opening for lower particle shedding
  • Full face opening
  • Ties with loops for adjustable fit
  • One size fits most
  • Availiable in non-sterile, clean-processed, and sterile versions
  • Available in bulk packaging with double transparent poly liners
  • Clean-processed garments meet ISO Class Cleanroom 4 standards
  • Sterilzed hoods = gamma sterilzed to SAL of 10-6
Specifications
Product TypeFull Face Opening
SizeUniversal
MaterialTyvek
ColorWhite
Closure TypeTies
DisposableSingle-use
ModelIC668B
Seam TypeBound
SterilitySterile
Quantity100/Cs.
IncludesTies with loops for fit
Hood TypeClean and Sterile
Cleanroom ClassClass 1-10 (ISO 3-4)
Unit SizeCase