Tyvek™ IsoClean™ Sleeve
Actual product may vary
DuPont
Tyvek™ IsoClean™ Sleeve
Comfortable, lightweight and durable.
Have Questions?
Catalog NumberPackaging
FIS17-988-107Bulk Packaged
Catalog number FIS17-988-107
Price (SGD)
450.00
Case
Packaging:
Bulk Packaged
Price (SGD)
450.00
Case
Comfortable, lightweight, and durable Tyvek sleeves are available in bulk or clean packaging.
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Elastic at both ends (bicep and wrist) for arm protection
  • 18 in. long
  • One size fits most
  • Bulk packaged in double transparent poly liners
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Product TypeControlled Environment Sleeve
SizeStandard
MaterialTyvek
ColorWhite
SterilityNon-sterile
DisposableSingle-use
Closure TypeElastic Ends
Seam TypeBound
Cuff StyleElastic
ModelIC501B
Length (English)18 in.
Length (Metric)45.7 cm
PackagingBulk Packaged
Unit SizeCase