360 Isolator Cleaning System
Actual product may vary
Berkshire
360 Isolator Cleaning System
Quick, easy and efficient cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeMaterialQuantity
FIS17-987-361Isolator Cleaning Tool CoverKnitted Polyester Elastic6/Pk.
BEREC360.ICT.1Isolater Cleaning ToolStainless Steel, Aluminum1 Ea.
BEREC360.ECST.20Isolator Cleaning Tool CoverKnitted Polyester Elastic10/Pk.
Catalog number FIS17-987-361
Price (SGD)
238.00
Pack
Product Type:
Isolator Cleaning Tool Cover
Material:
Knitted Polyester Elastic
Quantity:
6/Pk.
Price (SGD)
238.00
Pack
EasyClean 360 Isolator Cleaning Tool (17-987-360)
  • Durable, mirror finish, stainless-steel mop head swivels 360° for easy maneuverability
  • Head can be locked for 180° swivel
  • Triangular head shape allows for improved access in tight corners
  • Lightweight anodized aluminum handles (included) in two lengths: 14 in. (35cm) and 24 in. (61cm)
  • Quick-connect mechanism for easy handle attachment
  • Autoclavable (recommended maximum of 121°C for 30 min.)
  • Compatible for use in ISO Class 3/EU Grade A environments after sterilizing
EasyClean 360 ICT Sterile Knitted Polyester Elastic and Padded Covers (17-987-362)
  • Made from Berkshire's SuperPolx™ 1200A 100% continuous filament knitted polyester material
  • Laundered and packaged in an ISO Class 4 cleanroom for superior giver, particle and residue performance
  • Non-shedding and low-linting cover per USP requirements
  • Stretch fabric and elastic edge for easy on/off with secure fit on mop head
  • Absorbent polyester knit is abrasion resistant; handles a wide range of cleaning solutions
  • Compatible for use in ISO Class 3/EU Grade A Environments
  • Sterile
EasyClean 360 ICT Non-Sterile Knitted Polyester Elastic and Padded Covers (17-987-361)
  • Made from Berkshire's SuperPolx™ 1200A 100% continuous filament knitted polyester material
  • Laundered and packaged in an ISO Class 4 cleanroom for superior giver, particle and residue performance
  • Non-shedding and low-linting cover per USP requirements
  • Stretch fabric and elastic edge for easy on/off with secure fit on mop head
  • Absorbent polyester knit is abrasion resistant; handles a wide range of cleaning solutions
  • Compatible for use in ISO Class 3/EU Grade A Environments after sterilizing
  • Non-sterile
EasyClean 360 ICT Nonwoven Elastic Covers (17-987-363)
  • Made from Berkshire's Durx™ 670 55% cellulose/45% polyester nonwoven material
  • Absorbent rayon inner layer
  • Elastic edge allows easy installation and secure fit on mop head
  • Recommended for use in ISO Class 5-8/EU Grade C-D environments
Specifications
Product TypeIsolator Cleaning Tool Cover
ColorWhite
SterilityNon-sterile
AutoclavableAutoclavable
Quantity6/Pk.
Includes25 knitted polyester elastic covers and 1 knitted polyester padded cover/pack
For Use With (Equipment)EasyClean™ 360 Isolator Cleaning Tool
DescriptionMop Covers
For Use With (Application)Cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments.
Certifications/ComplianceISO Class 3/EU Grade A
MaterialKnitted Polyester Elastic
Unit SizePack