Gas Cylinder Brackets
Actual product may vary
First Safety
Gas Cylinder Brackets
All-steel construction wall mounted brackets
Have Questions?
Catalog NumberHoldsDimensions (L x W x H)Mounting
FIS17-987-155B2 Cylinders24 x 4 x 2 in.Wall
Catalog number FIS17-987-155B
Price (SGD)
265.00
Each
Holds:
2 Cylinders
Dimensions (L x W x H):
24 x 4 x 2 in.
Mounting:
Wall
Price (SGD)
265.00
Each
  • Straps rate 1138 psi and are the primary means of support, enabling cylinders to be held tight and secure against brackets
  • Support straps are 1.5 x 137cm (3.8 x 54 in.) long polypropylene with steel cinch buckle
  • Edges are protected with steel reinforced vinyl guarding to help maintain cylinders and provide extra grip
  • Steel parts are sealed with powder paint to assure long service life and chemical resistance
  • Optional Chain Sets (G-105) can be easily retrofit as a second means of support
  • Hold 10.16 to 30.48cm (4 to 12 in.) diameter cylinders
  • Constructed from 11 gauge hot rolled steel
Specifications
MaterialSteel
Height (Metric)5 cm
Height (English)2 in.
Dimensions (L x W x H)24 x 4 x 2 in.
Length (English)24 in.
Length (Metric)61 cm
Width (English)4 in.
Width (Metric)10.16 cm
Holds2 Cylinders
Gauge11g
IncludesSteel reinforced vinyl edge guarding, woven polypropylene straps, cinch style buckles, support strap
MountingWall
Product TypeBracket
Certifications/ComplianceNFPA1, CalOSHA, UFC
For Use With (Equipment)Gas cylinder
Weight (English)7 lb.
Weight (Metric)3.17 kg
Unit SizeEach