Safety Goggles Holder
Safety Goggles Holder
Actual product may vary
AK
Safety Goggles Holder
AK Safety Goggles Holder, PETG, Eyewear storage shelf, Description: Safety Glasses Holder, Device Holds: 12 pairs of glasses, For Use With: Stores glasses or goggles, Height: 12 in., 30.5cm, Includes: Hinged door, Length: 8 in., 20.3cm, Recommended Applications: Safety, Width: 17 in., 43.2cm
Have Questions?
Catalog number FIS17-987-140H
Price (SGD)
527.00
EA
Add to cart
Price (SGD)
527.00
EA
Add to cart
AK Safety Goggles Holder, PETG, Eyewear storage shelf, Description: Safety Glasses Holder, Device Holds: 12 pairs of glasses, For Use With: Stores glasses or goggles, Height: 12 in., 30.5cm, Includes: Hinged door, Length: 8 in., 20.3cm, Recommended Applications: Safety, Width: 17 in., 43.2cm
High-impact PETG unit holds up to 12 pairs of safety goggles. Hinged door keeps out dust and other contaminants.
Specifications
Height (English)12 in.
Height (Metric)30.5 cm
Width (English)17 in.
Width (Metric)43.2 cm
DescriptionSafety Glasses Holder
Device Holds12 pairs of glasses
Product TypeEyewear Storage Shelf
For Use With (Application)Stores glasses or goggles
IncludesHinged door
Length (English)8 in.
Length (Metric)20.3 cm
MaterialPETG
Unit SizeEA