AlphaTec™ 56-600 Urethane Aprons
Actual product may vary
Ansell
AlphaTec™ 56-600 Urethane Aprons
Urethane aprons designed for comfort and extended wear offers a combination of chemical protection, flexibility, and toughness.
Have Questions?
Catalog NumberLength (English)Length (Metric)
FIS17-987-113A48 in.121.92 cm
Catalog number FIS17-987-113A
Price (SGD)
49.00
Each
Length (English):
48 in.
Length (Metric):
121.92 cm
Price (SGD)
49.00
Each
  • Protect against oil, grease, and chemicals
  • Help wearers keep clean and protected from harmful contact
  • Urethane material offers superior flexibility and excellent resistance
  • Durable nylon reinforced to resist tearing, abrasion, and puncture yet remains flexible due to its lightweight construction
  • Sealed grommets allow for size adjustment without tear-out
  • Adjustable ties ensure optimal fit for a wide range of wearers
N/A
Specifications
TypeBib Apron
SizeMedium
MaterialUrethane
ColorYellow
DisposableReusable
Closure TypeIntegral Ties
SterilityNon-sterile
ConstructionNylon Reinforced Urethane, Sewn Edges
FinishSewn-edge
Latex FreeYes
No. of PocketsNone
ResistanceTearing, Abrasion, Puncture
Thickness7 mil
For Use With (Application)Food Service, Maintenance, Transferring Liquids
Dimensions (L x W)48 x 35 in. (121.92 x 88.9 cm)
Length (English)48 in.
Length (Metric)121.92 cm
Width (English)35 in.
Width (Metric)88.9 cm
No. per Case48/Cs.
No. per Pack1 Ea.
Packaging TypeCase
Seam TypeSewn
Unit SizeEach