AlphaTec™ 56-512 Hycar Apron
Actual product may vary
Ansell
AlphaTec™ 56-512 Hycar Apron
Heavy-duty cold-resistant Hycar™ nitrile apron ideal for chemical splash protection, and puncture and abrasion resistance.
Have Questions?
Catalog NumberConstruction
FIS17-987-107BNitrile Blend, Sewn Edge Finish
Catalog number FIS17-987-107B
Price (SGD)
66.00
Each
Construction:
Nitrile Blend, Sewn Edge Finish
Price (SGD)
66.00
Each
  • Hycar blend: Chemical splash, cut, puncture, oil, and abrasion resistance
  • Poly/cotton reinforcement: Enhanced protection, added cleanliness
  • Sealed grommets with two-piece 48-inch ties: Adjustability without tear-out
N/A
Specifications
TypeApron
SizeUniversal
MaterialPVC, Nitrile
ColorBlack
DisposableReusable
Closure TypeIntegral Ties
SterilityNon-sterile
ConstructionNitrile Blend, Sewn Edge Finish
FinishSewn-edge
Latex FreeYes
No. of PocketsNone
ResistanceChemical Splash, Cut, Puncture, Oil, Abrasion
Thickness17 to 19 mil
For Use With (Application)Food Service, Maintenance, Transferring Liquids
Dimensions (L x W)45 x 35 in. (114.3 x 88.9 cm)
Length (English)45 in.
Length (Metric)114.3 cm
Width (English)35 in.
Width (Metric)88.9 cm
Weight (English)18 oz.
No. per Case12/Cs.
No. per Pack1 Ea.
Seam TypeSewn
Unit SizeEach