Sterile Bacteriostatic Water Preservative
Actual product may vary
Haws
Sterile Bacteriostatic Water Preservative
Model 9082, 'sterile' bacteriostatic preservative for use in portable eyewash stations. One 5 oz. (148 ml) bottle will preserve 5 to 20 gallons (18.9 to 75.7 L) of potable water for up to three months. Four bottles per carton.
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS17-987-10Portable Eyewash Station
Catalog number FIS17-987-10
Price (SGD)
262.00
Case
For Use With (Equipment):
Portable Eyewash Station
Price (SGD)
262.00
Case
  • Model 9082, 'sterile' bacteriostatic preservative for use in portable eyewash stations.
  • One 8 oz. (237 ml) bottle will preserve 5 to 20 gallons (18.9 to 75.7 L) of potable water for up to 120 days.
  • This sterile solution is in compliance with FDA regulations.
  • Temperature storage requirements; store at 36-104°F, and use at 59-86°F.
  • Shelf life of 36 months
  • Four bottles per carton.
Specifications
Capacity (English)8 oz.
Capacity (Metric)237 mL
For Use With (Equipment)Portable Eyewash Station
Product TypeBacteriostatic Water Preservative
Shelf Life36 Months
Unit SizeCase