Flammables Undercounter Storage Cabinet
Actual product may vary
Justrite
Flammables Undercounter Storage Cabinet
Helps to keep work area safely organized
Have Questions?
Catalog NumberColor
FIS17-985-61AYellow
Catalog number FIS17-985-61A
Price (SGD)
-
Color:
Yellow
  • Double-wall construction provides 1-1/2 in. of insulating air space to help prevent the spread of flames High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • Two self-latching manual doors
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • SpillSlope safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Dual vents with flame arresters in back walls
  • Kickplate (3.5 x 2.5 in. [8.9 x 6.4cm]) mounts flush with other cabinetry
  • Leveling feet adjust to 1.13 in. (2.9cm) for alignment on uneven floors
  • Capacity: 22 gal. (83 L)
  • Backed by an exclusive 10-year warranty
  • Meet OSHA and NFPA requirements
Specifications
Certifications/ComplianceFM, NFPA, OSHA
Depth (English) Interior18.75 in.
Depth (Metric) Exterior55.9 cm
Depth (Metric) Interior47.6 cm
Dimensions (D x W x H) Interior18.75 x 31.75 x 27.37 in. (47.6 x 80.6 x 69.5 cm)
Door StyleManual-close
For Use With (Application)Flammables
Height (English) Interior27.37 in.
Height (Metric) Exterior88.9 cm
Height (Metric) Interior69.5 cm
IncludesOne adjustable shelves and mounting hardware, U-Loc™ handles with two keys, Haz-Alert™ reflective trilingual exterior labeling and Galvanized Steel SpillSlope™ shelves that direct all spills to the 5.1
No. of Doors2
No. of Shelves1
Surface CoatingEpoxy, Polyester
Weight (English)189.59 lb.
Weight (Metric)86 kg
Width (English) Interior31.75 in.
Width (Metric) Exterior88.9 cm
Width (Metric) Interior80.6 cm
ColorYellow
Depth (English) Exterior22 in.
Height (English) Exterior35 in.
MaterialSteel (CR)
Product TypeUndercounter safety cabinet
Width (English) Exterior35 in.
Unit SizeEach