Specifications
Product TypeGeneral Purpose Coveralls
SizeX-Large
GenderUnisex
MaterialSMS Fabric
ColorWhite
DisposableLimited-use
Ankle StyleElastic
Cuff StyleElastic
Neck StyleHooded
Closure TypeFront Zipper
SterilityNon-sterile
ResistanceDust
Back StyleElastic
Certifications/ComplianceNFPA 99
DescriptionMicroforce™ Barrier SMS Fabric
ModelKleenGuard™ A20
For Use With (Application)Laboratories, Decontamination Modesty Garment, Remediation, Pesticides, Heavy Dust Environments
Unit SizeCase