▸
Primary Antibodies
▸
All Primary Antibodies
▸
LYZ Mouse anti-Human, Clone: OTI1C9, lyophilized, TrueMAB™
Actual product may vary
DuPont
LYZ Mouse anti-Human, Clone: OTI1C9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Have Questions?
Catalog Number
Quantity
FIS17-685-211
100 μg
Catalog number
FIS17-685-211
Price (SGD)
1,634.00
Case
Add to cart
Quantity:
100 μg
Price (SGD)
1,634.00
Case
Add to cart
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Lysozyme is synthesized predomitly in reactive histiocytes rather than in resting, unstimulated phagocytes.
Specifications
Antigen
LYZ
Applications
Immunohistochemistry (Paraffin), Western Blot
Classification
Monoclonal
Clone
OTI1C9
Conjugate
Unconjugated
Formulation
PBS with 8% trehalose and no preservative; pH 7.3
Gene
LYZ
Gene Accession No.
P61626
Gene Alias
LYZF1, LZM
Gene Symbols
LYZ
Host Species
Mouse
Immunogen
Full length human recombit protein of human LYZ produced in HEK293T cell.
Purification Method
Affinity Chromatography
Quantity
100 μg
Regulatory Status
RUO
Primary or Secondary
Primary
Gene ID (Entrez)
4069
Target Species
Human
Product Type
Antibody
Form
Lyophilized
Isotype
IgG2b
Unit Size
Case
Contents & Storage
-20° C, Avoid Freeze/Thaw Cycles