Tyvek™ IsoClean™ Series 105 White Coveralls
Actual product may vary
Tyvek™ IsoClean™ Series 105 White Coveralls
Tyvek™ IsoClean™ Series 105 White Coveralls
Tyvek™ IsoClean™ Series 105 White Coveralls
Tyvek™ IsoClean™ Series 105 White Coveralls
Tyvek™ IsoClean™ Series 105 White Coveralls
Tyvek™ IsoClean™ Series 105 White Coveralls
Tyvek™ IsoClean™ Series 105 White Coveralls
Tyvek™ IsoClean™ Series 105 White Coveralls
Tyvek™ IsoClean™ Series 105 White Coveralls
DuPont
Tyvek™ IsoClean™ Series 105 White Coveralls
Ideal for use in controlled environments to protect against particulates and other contaminants.
Have Questions?
Catalog NumberSizeHeight (English)Height (Metric)
FIS17-685-210Medium5.5 to 5.10 ft.167.64 to 155.44 cm
Catalog number FIS17-685-210
Price (SGD)
1,634.00
Case
Size:
Medium
Height (English):
5.5 to 5.10 ft.
Height (Metric):
167.64 to 155.44 cm
Price (SGD)
1,634.00
Case
  • Light and durable coveralls provide coverage, including feet, while staying comfortable.
  • Use in controlled environments with high standards for particle and microbiological contamination control.
  • Clean processed and sterile
  • Spinning process creates barrier to particles, micro-organisms, and non-hazardous liquids
  • Lightweight, comfortable and durable
  • Lowest level particle shedding
  • Recyclable
  • Serged seams
  • Standard hood, elastic opening
  • Elastic wrists and ankles
  • Attached thumb loops
  • Zipper closure
  • Attached boots, PVC sole
  • Pharmaceutical, biotech, medical device, electronic settings
Specifications
Product TypeCleanroom Coveralls
SizeMedium
GenderUnisex
MaterialTyvek
ColorWhite
DisposableSingle-use
Waist StyleNon-elastic
Ankle StyleElastic, Attached Boots
Cuff StyleThumb Loop
Neck StyleHooded
Hood TypeElastic
Closure TypeZipper
Seam TypeSerged
SterilitySterile
ResistanceParticle, Non-toxic Particulates
DescriptionElastic wrist and ankle, attached thumb loops, standard hood with elastic opening, attached boot with PVC soles, set sleeves
ModelIC105SWH
Size (English) Chest36 to 38 in.
Size (English) Inseam28 in.
Size (Metric) Chest91.44 to 96.52 cm
Size (Metric) Inseam71 cm
Thickness5 mil
For Use With (Application)Barrier to dry particles, microorganisms, and non-hazardous liquids
Length (English) Sleeve33.25 in.
Length (Metric) Sleeve84.45 cm
Height (English)5.5 to 5.10 ft.
Height (Metric)167.64 to 155.44 cm
Cleanroom ClassClass 100-100,000 (ISO 5-8)
Unit SizeCase