▸
Facility Safety and Maintenance
▸
Hand Tools and Power Tools
▸
Miscellaneous Tools
▸
Roto-Pic Vacuum System Tips
Actual product may vary
Excelta
Roto-Pic Vacuum System Tips
For handling small parts
Have Questions?
Catalog Number
Product Type
Diameter (Metric) Inside
Material
FIS17-468-544
Roto-pic Tip
0.015 cm
PTFE
Catalog number
FIS17-468-544
Price (SGD)
238.00
Each
Add to cart
Product Type:
Roto-pic Tip
Diameter (Metric) Inside:
0.015 cm
Material:
PTFE
Price (SGD)
238.00
Each
Add to cart
Roto-Pic™ vacuum straight tips are for handling very small parts.
Specifications
Product Type
Roto-pic Tip
For Use With (Equipment)
Hand piece and probes
Material
PTFE
Diameter (English) Inside
0.006 in.
Diameter (Metric) Inside
0.015 cm
For Use With (Application)
Handling small parts
Includes
0.006 in. (0.15mm) PTFE Tip