Roto-Pic™ Vacuum Handpiece
Actual product may vary
Excelta
Roto-Pic™ Vacuum Handpiece
Five star replacement handpiece used with Roto-Pic vacuum system
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS17-468-531Vacuum System, Ultra-precision
Catalog number FIS17-468-531
Price (SGD)
1,069.00
Each
For Use With (Equipment):
Vacuum System, Ultra-precision
Price (SGD)
1,069.00
Each
The 2000A is the Roto-pic™ replacement hand piece. The hand piece includes a slide switch for on-off and a rotary wheel to turn the very tip for perfect alignment of the part.
Specifications
Product TypeRoto-pic Vacuum Hand Piece
For Use With (Equipment)Vacuum System, Ultra-precision
For Use With (Application)For vacuum pickup of chips and wafers
IncludesPositive hold-and-release slide bar, 360deg. rotatable round nose wheel, replaceable tips and silicon rubber cups
Unit SizeEach