Self-Opening Micro Scissors
Actual product may vary
Excelta
Self-Opening Micro Scissors
Meets Rockwell C-50 hardness test scale
Have Questions?
Catalog NumberBlade Style
FIS17-467-497Angled at 45°
Catalog number FIS17-467-497
Price (SGD)
123.00
Each
Blade Style:
Angled at 45°
Price (SGD)
123.00
Each
  • Fine precision
  • Made from stainless-steel
  • Overall length: 3 in. (7.62cm)
Specifications
Blade StyleAngled at 45°
Length (English)3 in.
Length (Metric)7.62 cm
MaterialStainless Steel
DescriptionHardened to Rockwell C-50
Unit SizeEach