Electronic Style Tweezers With Anti-Crush Feature
Actual product may vary
Electronic Style Tweezers With Anti-Crush Feature
Electronic Style Tweezers With Anti-Crush Feature
Excelta
Electronic Style Tweezers With Anti-Crush Feature
For general assembly and light magnification
Have Questions?
Catalog NumberLength (Metric)
FIS17-467-270120 mm
Catalog number FIS17-467-270
Price (SGD)
203.00
Each
Length (Metric):
120 mm
Price (SGD)
203.00
Each
  • High Precision
  • NEVERUST™ stainless steel
Specifications
Product TypeTweezers, Electronic
Length (English)4.75 in.
Length (Metric)120 mm
MaterialStainless Steel, Neverrust™
Height (English)0.015 in.
Height (Metric)0.038 cm
QuantityEach
Tip StyleFlat, Round
Width (English)0.32 in.
Width (Metric)0.81 cm
Unit SizeEach