Small Parts Handling Tweezers
Actual product may vary
Excelta
Small Parts Handling Tweezers
Choice of small, round, broad angle, broad and curved flat tips
Have Questions?
Catalog NumberTip Style
FIS17-467-254Offset Broad Flat Tip
Catalog number FIS17-467-254
Price (SGD)
34.00
Each
Tip Style:
Offset Broad Flat Tip
Price (SGD)
34.00
Each
  • Available in high and general precision
  • Made of carbon tool steel, 300 and 400 series stainless-steel
  • Choice of small, round, broad angle, broad and curved flat tips
Specifications
Product TypeTweezers
Length (English)4.75 in.
Length (Metric)120 mm
MaterialStainless Steel
For Use With (Application)Ideal for picking flat objects off a flat surface, serrated handles for better control
Height (English)0.015 in.
Height (Metric)0.038 cm
QuantityEach
Tip StyleOffset Broad Flat Tip
Width (English)0.32 in.
Width (Metric)0.81 cm
Unit SizeEach