General-Purpose Tweezers
Actual product may vary
General-Purpose Tweezers
General-Purpose Tweezers
Excelta
General-Purpose Tweezers
Feature serrated tips, finger serrations for better control and very strong points
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Tip Style
FIS17-467-242203 mmLong Angled Blunt
FIS17-467-238152 mmBroad
Catalog number FIS17-467-242
Price (SGD)
41.00
Each
Length (Metric):
203 mm
Tip Style:
Long Angled Blunt
Price (SGD)
41.00
Each
  • Stainless steel
  • Many styles offer acid resistance
  • Angled with serrated broad point
Specifications
Product TypeTweezers, General Purpose
Length (English)8 in.
Length (Metric)203 mm
MaterialStainless Steel
QuantityEach
Tip StyleLong Angled Blunt
Width (English)0.46 in.
Width (Metric)1.168 cm
Unit SizeEach