General-Purpose Tweezers with Angle Tips with Medium Fine Serrated Points
Actual product may vary
Excelta
General-Purpose Tweezers with Angle Tips with Medium Fine Serrated Points
Alignment pin to keeps tips aligned during high strain applications
Have Questions?
Catalog NumberMaterial
FIS17-467-237Stainless Steel, Neverrust™
Catalog number FIS17-467-237
Price (SGD)
248.00
Each
Material:
Stainless Steel, Neverrust™
Price (SGD)
248.00
Each
  • Double cut inside serrations provide superior gripping when handling small parts
  • Outside finger serrations for easy handling
  • Length: 5.5 in. (14cm)
Specifications
Length (English)5.5 in.
Length (Metric)140 mm
MaterialStainless Steel, Neverrust™
QuantityEach
Tip StyleAngled
Width (English)0.3 in.
Width (Metric)0.81 cm
Product TypeTweezers, General Purpose
Unit SizeEach