Precision Tweezers with Tapered Tips with High Precision Points
Actual product may vary
Excelta
Precision Tweezers with Tapered Tips with High Precision Points
Precision Tweezers with Tapered Tips with High Precision Points offer extreme precision for work under microscopes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial
FIS17-467-166Stainless Steel
FIS17-467-165Stainless Steel
ECT5-SStainless Steel
Catalog number FIS17-467-166
Price (SGD)
23.00
Each
Material:
Stainless Steel
Price (SGD)
23.00
Each
  • Antimagnetic stainless steel, Stainless steel or Carbon steel
  • Straight Tapered ultra-fine tip enhances visibility under microscopes
  • Overall length: 4.25 in.
Specifications
Product TypeTweezers, Electronic
Length (English)4.25 in.
Length (Metric)108 mm
MaterialStainless Steel
Height (English)0.015 in.
Height (Metric)0.038 cm
Quantity1 Ea.
StyleHigh-precision
Tip StyleStraight Tapered, Ultra Fine
Width (English)0.32 in.
Width (Metric)0.812 cm
Unit SizeEach