Reverse-Action Tweezers
Actual product may vary
Excelta
Reverse-Action Tweezers
Tweezers designed to hold objects without finger strain
Have Questions?
Catalog NumberLength (English)Tip Style
FIS17-447-1084.75 in.Tapered, Flat Round
Catalog number FIS17-447-108
Price (SGD)
56.00
Each
Length (English):
4.75 in.
Tip Style:
Tapered, Flat Round
Price (SGD)
56.00
Each
  • AISI 303 anti-magnetic, high-precision stainless-steel
  • Closed at rest, press to open
  • Ideal for holding any small items for extended periods of time
Specifications
Length (English)4.75 in.
Length (Metric)120 mm
MaterialStainless Steel
For Use With (Application)Holding any small items for extended periods of time
Height (English)1 in.
Height (Metric)2.54 cm
QuantityEach
Tip StyleTapered, Flat Round
Width (English)0.32 in.
Width (Metric)0.81 cm
Product TypeTweezers, Reverse Action
Unit SizeEach