North™ Protecto-Shield Face Shields
Actual product may vary
Splash Shield
North™ Protecto-Shield Face Shields
Fits well over glasses and protective spectacles
Have Questions?
Catalog number FIS17-310
Price (SGD)
134.00
Pack
Price (SGD)
134.00
Pack
Fits well over glasses and protective spectacles
  • Disposable
  • Constructed from high-quality optically clear, distortion-free 7mil polyester film
  • Headpiece is held in place by stretch strap
Specifications
Thickness (English)0.007 in.
Thickness (Metric)0.1778 mm
Certifications/ComplianceOSHA 29 CFR Part 1910.1030, CDC recommended for protective eyewear
Lens TintClear
Product TypeSplash Shield
For Use With (Application)Fits well over glasses and protective spectacles
MaterialPolyester