Kimwipes™ Lens Cleaning Stations
Kimwipes™ Lens Cleaning Stations
Actual product may vary
Kimwipes™ Lens Cleaning Stations
Use anywhere glasses or safety lenses are worn.
Have Questions?
Catalog NumberDescription
FIS17-228BLens Cleaning Station
Catalog number FIS17-228B
Price (SGD)
-
Description:
Lens Cleaning Station
Use anywhere glasses or safety lenses are worn
  • Handy one-at-a-time 4.50 × 8.50 in. (11 × 22cm) wipers won't scratch glass or plastic lenses when used wet
  • Silicone-free, antistatic, antifog cleaning solution removes dirt quickly and easily
Specifications
Dimensions (L x W)4.4 x 8.4 in. (11.1 x 21.3 cm)
DescriptionLens Cleaning Station
Product TypeCleaning station
For Use With (Application)nonscratching glass or plastic lens wipes
Includes1120 wipes and one 16 oz. cleaner
Unit SizeEA