SPILL-X-FP Formaldehyde Treatment Kit
Actual product may vary
Ansul
SPILL-X-FP Formaldehyde Treatment Kit
Free-flowing dry SPILL-X-FP agent to treat spills
Have Questions?
Catalog NumberPackaging Quantity
FIS17-200-66 Bottles/Pk.
Catalog number FIS17-200-6
Price (SGD)
-
Packaging Quantity:
6 Bottles/Pk.
Free-flowing dry SPILL-X-FP agent to treat spills
SPILL-X-FP agent ihas been developed to reach with all formaldehyde solutions. The reaction of SPILL-X-FP agent with aqueous formaldehyde is both temperature and concentration dependent with respect to formaldehyde solutions strenght. Agent components have been formulate to regulate the reaction at an acidic pH, yielding a positive effect on reaction rate.
Specifications
BaseUrea
ColorGreen
Description0.80 g/mL density, 2.1 pH
TypeFormaldehyde Spill Control Treatment
For Use With (Application)Formaldehyde spills
Packaging Quantity6 Bottles/Pk.
Unit SizePack