Portable Eyewashes
Actual product may vary
Haws
Portable Eyewashes
Ideal for remote locations
Have Questions?
Catalog NumberCapacity (English)
FIS17-163-12048 oz.
Catalog number FIS17-163-1
Price (SGD)
1,516.00
Each
Capacity (English):
2048 oz.
Price (SGD)
1,516.00
Each
Portable eyewashes serve as ANSI-approved eyewash units in remote locations—anywhere plumbed water is not available. Choose from gravity-operated or air pressure-operated models. Both units come with one 8 oz. (237mL) bottle of bacteriostatic preservative that permits water storage in the units for up to six months.
Self-Contained, Gravity-Operated Eyewash Station.
  • Unit exceeds the ANSI standard with a 16 gal. (60.6L) capacity for more than 15 minutes of flushing
  • Its Safety Green high-density polyethylene tank is highly visible and easy to transport
  • Built-in top handle and two integral side handles facilitate two-person handling when filled
  • Wide fill opening allows easy filling, inspection, and cleaning
  • A threaded cap prevents insects from entering tank; recessed area encloses eyewash heads, protecting them from debris
  • To activate heads, grasp the arm containing the heads and simply pull down
  • Store on a shelf or mount on a wall with the included bracket
  • Comes with a drain hose for emptying ease
Air Pressure-Operated Eyewash.
  • Consists of ABS plastic spray heads with dust covers, with proprietary Soft-Flo™ antisurge design, mounted on a 10 gal. (38L) stainless-steel tank with carrying handles
  • Stainless-steel push flag activates the chrome-plated brass ball valve, which stays open until manually closed Protective dust covers on the spray heads dislodge when the unit is activated
  • The tank delivers a 15-minute full flow eyewash or a 2-minute body spray via stored air pressure
  • The tank is outfitted with tire-type air valve (recommended pressure: 87 psi), pressure gauge, safety relief valve (set at 110 psi), and additional relief valve built into tank cover
  • Optional 6 ft. long (1.8m) body spray with a heavy-duty rubber hose, squeeze lever handle, valve, and ABS plastic Feather-Flo™ spray head sold separately
Specifications
Capacity (English)2048 oz.
Capacity (Metric)60566.58 mL
Certifications/ComplianceANSI Z358.1-2009, FDA
ColorGreen
Depth (English)19.5 in.
Depth (Metric)49.53 cm
Flow Rate0.4 gpm
Height (English)22.5 in.
Height (Metric)57.15 cm
IncludesPreservative, Drain Hose, Brackets, Built-in Top, Side Handle
MaterialPolyethylene
MountingWall or Cart
Product TypeEyewash Station
Weight (English)140 lb.
Weight (Metric)63.50 kg
Width (English)21.5 in.
Width (Metric)54.61 cm
Unit SizeEach