Flammable Liquid Safety Storage Cabinet
Actual product may vary
Eagle
Flammable Liquid Safety Storage Cabinet
Save time and money by keeping normally dangerous items on hand. With Eagle's Flammable Liquid Safety Storage Cabinet, you can safely keep flammable liquids near work areas.
Have Questions?
Catalog NumberLoad Bearing CapacityCertifications/ComplianceNo. of DoorsNo. of Shelves
FIS17-156F60 gal.OSHA, NFPA22
Catalog number FIS17-156F
Price (SGD)
6,023.00
Each
Load Bearing Capacity:
60 gal.
Certifications/Compliance:
OSHA, NFPA
No. of Doors:
2
No. of Shelves:
2
Price (SGD)
6,023.00
Each
Save time and money by keeping normally dangerous items on hand. With Eagle's Flammable Liquid Safety Storage Cabinet, you can safely keep flammable liquids near work areas.
  • Top and bottom vents, with baffle and cap, fitted with 2 in. (5cm) pipe thread on sidewalls for outside venting
  • Double walls of 18-gauge steel provide 1.5 in. (3.8cm) air space
  • 2 in. (5cm) raised sill contains spills
  • Three-point key lock, 2 keys
  • Manually operated doors
  • Four adjustable leveling feet
  • Adjustable galvanized steel shelves supported by four brackets hold over 500 lb. (227kg) each Labeled FLAMMABLE—KEEP FIRE AWAY
Door Choices
  • Available with one or two hinged doors or with bifold door, hinged left, with piano hinge in the middle (2, 4, 12, and 16 gal. units have one door)
Specifications
Certifications/ComplianceOSHA, NFPA
Depth (English) Interior30.34 in.
Depth (Metric) Exterior86 cm
Depth (Metric) Interior77.1 cm
Dimensions (D x W x H)31.25 x 31.25 x 65 in.
Dimensions (D x W x H) Interior30.34 x 30.75 x 60.5 in.
Door StyleManual-close
For Use With (Application)Flammables
Height (English) Interior60.5 in.
Height (Metric) Exterior165.1 cm
Height (Metric) Interior153.7 cm
IncludesGrounding attachments (top and bottom vents, with baffle and cap, fitted with 2 in. pipe thread on sidewalls), three-point key lock on doors, 5cm raised, leakproof door sill contains spills and four leveling feet, flammable-keep fire away label, 2 keys
Load Bearing Capacity60 gal.
No. of Doors2
No. of Shelves2
Weight (English)322 lb.
Weight (Metric)146.05 kg
Width (English) Interior30.75 in.
Width (Metric) Exterior86 cm
Width (Metric) Interior78.1 cm
ColorYellow
Depth (English) Exterior34 in.
Height (English) Exterior65 in.
MaterialSteel
Product TypeFlammables Storage Cabinet (Floor Model)
Width (English) Exterior34 in.
Unit SizeEach