Indoor Drum Storage Cabinet
Indoor Drum Storage Cabinet
Actual product may vary
Justrite
Indoor Drum Storage Cabinet
Justrite's Indoor Drum Storage Cabinet features a removable, adjustable, half-depth shelf with enough headspace to accommodate a drum with a pump or funnel. This means the cabinet can be used as a pumping or waste accumulation station , storage for Class III materials, or for stowing of spare accessories.
Have Questions?
Catalog NumberDimensions (D x W x H) InteriorWidth (Metric) ExteriorWeight (Metric)Certifications/Compliance
FIS17-150B30.34 x 56 x 60.5 in.149.9 cm277 kgNFPA, OSHA
Catalog number FIS17-150B
Price (SGD)
-
Dimensions (D x W x H) Interior:
30.34 x 56 x 60.5 in.
Width (Metric) Exterior:
149.9 cm
Weight (Metric):
277 kg
Certifications/Compliance:
NFPA, OSHA
  • Double-wall 18-gauge steel with 1.5 in. (3.8cm) insulating air space
  • Three-point self-latching manual doors with double key set
  • Dual 2 in. (5cm) vents with flame arresters
  • Spill-catcher shelf adjustable on 2.25 in. (5.7cm) centers
  • Adjustable leveling feet
  • High-visibility warning label
  • 2 in. (5cm) leakproof sill to capture spills
  • Welded for superior strength
  • Powder-paint finish for maximum durability and chemical resistance
Specifications
Certifications/ComplianceNFPA, OSHA
Depth (English) Interior30.34 in.
Depth (Metric) Exterior86.4 cm
Depth (Metric) Interior77.1 cm
Dimensions (D x W x H) Interior30.34 x 56 x 60.5 in.
Door StyleManual-close
For Use With (Application)Flammables
Height (English) Interior60.5 in.
Height (Metric) Exterior165.1 cm
Height (Metric) Interior153.7 cm
IncludesHalf-depth shelf, installed pumps, drum support, level adjusting feet and hazardous warning label
Load Bearing Capacity110 gal.
No. of Doors2
No. of Shelves1
Weight (English)610 lb.
Weight (Metric)277 kg
Width (English) Interior56 in.
Width (Metric) Exterior149.9 cm
Width (Metric) Interior142.2 cm
ColorYellow
Depth (English) Exterior34 in.
Height (English) Exterior65 in.
MaterialSteel (CR)
Product TypeVertical drum storage cabinet
Width (English) Exterior59 in.
Unit SizeEA