Search Fisher Scientific
| Catalog Number | Size | Length (English) |
|---|---|---|
| FIS17-100-876A | X-Large | 41 in. |
| FIS17-100-873A | Small | 35 in. |
| FIS17-100-875A | Large | 39 in. |
Note:
Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Polypropylene Lab Coats with Elastic Cuffs, and Fisherbrand Basic Protection Disposable Lab Coats with Pockets. SMS coats are also available in White, Teal, Purple, Cranberry and Blueberry.