NAIF1 Polyclonal Antibody
Actual product may vary
NAIF1 Polyclonal Antibody
NAIF1 Polyclonal Antibody
NAIF1 Polyclonal Antibody
NAIF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Have Questions?
Catalog NumberQuantity
FIS17-100-844100 μg
Catalog number FIS17-100-844
Price (SGD)
-
Quantity:
100 μg
NAIF1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Human NAIF1 is located on chromosome 9q34.11 and encodes 327 amino acids with a homeodomain-like region and two nuclear localization signals at its N-terminal region. NAIF1 is conserved across diverse species, including human, mouse, crab-eating macaque, dog, chicken and frog, and shares no obvious homology to any known genes or proteins. NAIF1 was predominantly localized in the nucleus. Overexpression of NAIF1 inhibited cell growth and induced apoptosis. Furthermore, NAIF1 transfection caused both decreases in mitochondrial membrane potential and caspase-3 activation. In summary, NAIF1 is a nuclear protein that induces apoptosis when overexpressed.
Specifications
AntigenNAIF1
ApplicationsELISA, Immunohistochemistry (Paraffin), Western Blot
ClassificationPolyclonal
Concentration3 mg/mL
ConjugateUnconjugated
FormulationPBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
GeneNAIF1
Gene Accession No.Q69YI7, Q6PFD7
Gene Alias2310007O20Rik; 4933440H19Rik; bA379C10.2; C9orf90; NAIF1; nuclear apoptosis inducing factor 1; nuclear apoptosis-inducing factor 1
Gene SymbolsNAIF1
Host SpeciesRabbit
ImmunogenRecombinant Human Nuclear apoptosis-inducing factor 1 protein (1-172AA).
Purification MethodProtein G
Quantity100 μg
Regulatory StatusRUO
Primary or SecondaryPrimary
Gene ID (Entrez)203245, 71254
Target SpeciesHuman, Mouse
Product TypeAntibody
FormLiquid
IsotypeIgG
Unit SizePack
Contents & Storage
-20°C or -80°C if preferred